Learn More
Falcon™ Pipet Controller Hydrophobic PTFE Filters
GREENER_CHOICE
Product Code. 15339840 Shop All Corning Products
Change view
Click to view available options
Pore Size:
0.2 μm
0.45 μm
Unit Size:
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size unitSize
15339840 0.45 μm Pack of 5
15329840 0.2 μm Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15339840 Supplier Falcon™ Supplier No. 357473
In Stock
Add to basket
Add to basket
Replacement hydrophobic PTFE disc filters
Additional filters are available as standard accessories. The filters, pipet holder, and nose piece are autoclavable.
TRUSTED_SUSTAINABILITY
Specifications
Material PTFE
Pore Size 0.45 μm
For Use With (Equipment) For use with pipet controller
Quantity 5/Cs.