Learn More
Falcon™ Vacutainer Lid
GREENER_CHOICE
Product Code. 10308372 Shop All Corning Products
Click to view available options
:
For 4.5 oz. or 8 oz. (125 or 220mL) Vacutainers
Product Code. 10308372
Brand: Falcon™ 354017
For sterile sample containers
Lid for Falcon™ Sterile Sample Containers. Use with either 4.5oz or 8oz size containers (Cat. #s 354013, 354014, 354015, and 354020). Sterile, 20/bag, 500/case
TRUSTED_SUSTAINABILITY
Specifications
Material Polypropylene
No. per Pack 20/Pk.
No. per Case 500/Cs.
Sterility Sterile
For Use With (Equipment) 4.5 or 8 oz. Sample Containers