missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FALDH Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99237-50ul
This item is not returnable.
View return policy
Description
FALDH Polyclonal antibody specifically detects FALDH in Mouse samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| FALDH | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:500-1:2000 | |
| Aldehyde dehydrogenase 10, aldehyde dehydrogenase 3 family, member A2, Aldehyde dehydrogenase family 3 member A2, ALDH10FLJ20851, EC 1.2.1, EC 1.2.1.3, FALDHDKFZp686E23276, fatty aldehyde dehydrogenase, Microsomal aldehyde dehydrogenase, SLS | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Mouse FALDH. | |
| 50 μg | |
| Stem Cell Markers | |
| 224 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction