missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FALDH Antibody (OTI2A7), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70159IR
This item is not returnable.
View return policy
Description
FALDH Monoclonal specifically detects FALDH in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
Specifications
| FALDH | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ALDH3A2 | |
| Full length recombinant protein of human FALDH (NP_001026976) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Rat | |
| IgG2b |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| OTI2A7 | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| Aldehyde dehydrogenase 10, aldehyde dehydrogenase 3 family, member A2, Aldehyde dehydrogenase family 3 member A2, ALDH10FLJ20851, EC 1.2.1, EC 1.2.1.3, FALDHDKFZp686E23276, fatty aldehyde dehydrogenase, Microsomal aldehyde dehydrogenase, SLS | |
| Mouse | |
| Affinity Purified | |
| Stem Cell Markers | |
| 224.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction