missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM101B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570891
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Dog: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rat: 100%.
RFLNB is a protein coding gene involved in the regulation of the perinuclear actin network and nuclear shape through interaction with filamins. Plays an essential role in the formation of cartilaginous skeletal elements.
Specifications
| FAM101B | |
| Polyclonal | |
| Unconjugated | |
| RFLNB | |
| 1500005K14Rik; cfm; Cfm1; Fam101b; family with sequence similarity 101 member B; family with sequence similarity 101, member B; filamin-interacting protein FAM101B; filamin-interacting protein FAM101B; refilin-B; MGC45871; protein FAM101B; refilin B; RefilinB; refilin-B; regulator of filamin protein B; RFLNB; RGD1359691 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 359845 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8N5W9 | |
| RFLNB | |
| synthetic peptide directed towards the N-terminal region of human FAM101B. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction