missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FAM101B Polyclonal Antibody
GREENER_CHOICE

Product Code. 16361405
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16361405 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16361405 Supplier Invitrogen™ Supplier No. PA584455

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: EGVEFDPLPP KEVRYTSLVK YDSERHFIDD VQLPLGLAVA SCSQTVTCVP Highest antigen sequence indentity to the following orthologs: Mouse - 88%, Rat - 88%.

RFLNB is a protein coding gene involved in the regulation of the perinuclear actin network and nuclear shape through interaction with filamins. Plays an essential role in the formation of cartilaginous skeletal elements.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM101B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RFLNB
Gene Accession No. Q8N5W9
Gene Alias 1500005K14Rik; cfm; Cfm1; Fam101b; family with sequence similarity 101 member B; family with sequence similarity 101, member B; filamin-interacting protein FAM101B; filamin-interacting protein FAM101B; refilin-B; MGC45871; protein FAM101B; refilin B; RefilinB; refilin-B; regulator of filamin protein B; RFLNB; RGD1359691
Gene Symbols RFLNB
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RFLNB. Recombinant protein control fragment (Product #RP-108218).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 359845
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.