missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM104B Antibody (OTI1D10), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72313AF750
This item is not returnable.
View return policy
Description
FAM104B Monoclonal specifically detects FAM104B in Human samples. It is validated for Western Blot.
Specifications
| FAM104B | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| FAM104B | |
| Full length recombinant protein of human FAM104B (NP_612371) produced in E.coli. | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot | |
| OTI1D10 | |
| Western Blot | |
| chromosome X open reading frame 44, CXorf44, family with sequence similarity 104, member B, FLJ18375, FLJ20434, hypothetical protein LOC90736 | |
| Mouse | |
| 0.1 mL | |
| 90736 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction