missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ FAM105B/OTULIN Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30584
This item is not returnable.
View return policy
Description
FAM105B/OTULIN Polyclonal antibody specifically detects FAM105B/OTULIN in Human samples. It is validated for Immunoprecipitation
Specifications
| FAM105B/OTULIN | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 90268 | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Immunoprecipitation | |
| Unconjugated | |
| FAM105B family with sequence similarity 105, member B | |
| between 302 and 352 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction