missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM107A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97556-100ul
This item is not returnable.
View return policy
Description
FAM107A Polyclonal antibody specifically detects FAM107A in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| FAM107A | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:100-1:400 | |
| downregulated in renal cell carcinoma, DRR1family with sequence similarity 107 member A transcript, family with sequence similarity 107, member A, FLJ30158, FLJ45473, Protein TU3A, TU3ADown-regulated in renal cell carcinoma 1 | |
| Produced in rabbits immunized with E. coli-derived Human FAM107A fragment. | |
| 100 μg | |
| Cancer, Cell Biology, Cell Cycle and Replication | |
| 11170 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction