missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM107A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563200
This item is not returnable.
View return policy
Description
Immunogen sequence: NQLIKKKKEE LEAKRLQCPF EQELLRRQQR LNQLEKPPEK EEDHAPEFIK VRENLRRIAT LTSEER Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 91%.
FAM107A (Family With Sequence Similarity 107 Member A) is a Protein Coding gene. Diseases associated with FAM107A include Neuroblastoma and Brain Cancer. An important paralog of this gene isFAM107B.
Specifications
| FAM107A | |
| Polyclonal | |
| Unconjugated | |
| FAM107A | |
| Actin-associated protein FAM107A; actin-associated protein FAM107A; protein FAM107A; down-regulated in renal cell carcinoma 1; DRR1; FAM107A; family with sequence similarity 107 member A; family with sequence similarity 107, member A; protein FAM107A; Protein TU3A; RGD1306327; Tu3a | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11170 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O95990 | |
| FAM107A | |
| Recombinant protein corresponding to Human FAM107A. Recombinant protein control fragment (Product #RP-91874). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction