missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FAM107B Polyclonal Antibody
GREENER_CHOICE

Product Code. 16391865
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16391865 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16391865 Supplier Invitrogen™ Supplier No. PA584920

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: LHRELLMNQK RGLAPQNKPE LQKVMEKRKR DQVIKQKEEE AQKKKSDLEI ELLKRQQKLE QLELEKQ Highest antigen sequence indentity to the following orthologs: Rat - 99%, Mouse - 99%.

FAM107B is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM107B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM107B
Gene Accession No. Q9H098
Gene Alias 3110001A13Rik; AA589595; AA959793; AI132312; axoneme-associated protein mst101(2)-like; C10orf45; Fam107b; fam107b protein; family with sequence similarity 107 member B; family with sequence similarity 107, member B; FLJ45505; HITS; hypothetical protein LOC386641; hypothetical protein MGC11034-like protein; MGC11034; MGC137524 protein; novel DUF1151 domain containing protein; protein FAM107B; wu:fc52f02; zgc:56216
Gene Symbols FAM107B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM107B. Recombinant protein control fragment (Product #RP-108609).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83641
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.