missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM111B Antibody [Janelia Fluor™ 585], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-18411JF585
This item is not returnable.
View return policy
Description
FAM111B Polyclonal antibody specifically detects FAM111B in Human samples. It is validated for Western Blot
Specifications
| FAM111B | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 374393 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Janelia Fluor 585 | |
| family with sequence similarity 111, member B | |
| Recognizes region between residues Between between 225 and 275.. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction