missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM114A1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-57592-25ul
This item is not returnable.
View return policy
Description
FAM114A1 Polyclonal specifically detects FAM114A1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| FAM114A1 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 1-4 μg/mL | |
| family with sequence similarity 114, member A1, FLJ33151, nervous system over expressed protein 20, Nervous system over-expressed protein 20, NOXP20, Protein FAM114A1, protein NOXP20 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 92689 | |
| Human | |
| Purified |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| FAM114A1 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:TPDKLNKAMKRAHDWVEEDQTVVSVDVAKVSEEETKKEEKEEKSQDPQEDKKEEKKTKTIEEVYMSSIESLAE | |
| 25 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction