missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM119A Antibody (OTI2G7), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72423AF350
This item is not returnable.
View return policy
Description
FAM119A Monoclonal specifically detects FAM119A in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
Specifications
| FAM119A | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| METTL21A | |
| Full length recombinant protein of human FAM119A (NP_660323) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2b |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| OTI2G7 | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| EC 2.1.1.-, FAM119A, HCA557B, Hepatocellular carcinoma-associated antigen 557b, hypothetical protein LOC151194, LOC151194, member A, methyltransferase like 21A, MGC45373 | |
| Mouse | |
| Affinity Purified | |
| Proteases & Other Enzymes | |
| 151194.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction