missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM119B Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-26094AF700
This item is not returnable.
View return policy
Description
FAM119B Polyclonal antibody specifically detects FAM119B in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| FAM119B | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| DKFZP586D0919, EC 2.1.1.-, FAM119B, family with sequence similarity 119, member B, HCA557A, Hepatocellular carcinoma-associated antigen 557a, hepatocellularcarcinoma-associated antigen HCA557a, hypothetical protein LOC25895, methyltransferase like 21B | |
| Produced in rabbits immunized with E. coli-derived Human FAM119B fragment | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 25895 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction