missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM120A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531766
This item is not returnable.
View return policy
Description
Recommended positive controls: MCF-7. Predicted reactivity: Mouse (95%), Rat (93%), Bovine (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
May participate in mRNA transport in the cytoplasm. Critical component of the oxidative stress-induced survival signaling. Activates src family kinases and acts as a scaffolding protein enabling src family kinases to phosphorylate and activate PI3-kinase. Binds RNA and promotes the secretion of IGF-II. May play a pivotal role in the progression of scirrhous-type gastric cancer by supporting cancer cell survival in environments with various oxidative stresses. [UniProt].
Specifications
| FAM120A | |
| Polyclonal | |
| Unconjugated | |
| Fam120a | |
| C9orf10; Constitutive coactivator of PPAR-gamma-like protein 1; DNAPTP1; DNAPTP5; Fam120a; family with sequence similarity 120, member A; family with sequence similarity 120A; Kiaa0183; Ossa; oxidative stess-associated Src activator; oxidative stress-associated Src activator; Protein FAM120A; RGD1306643 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23196 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q9NZB2 | |
| Fam120a | |
| Recombinant protein encompassing a sequence within the center region of human FAM120A. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction