missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM120B Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76315-0.025mg
This item is not returnable.
View return policy
Description
FAM120B Polyclonal specifically detects FAM120B in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.
Specifications
| FAM120B | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| CCPGKIAA0183, Constitutive coactivator of PPARG, Constitutive coactivator of PPAR-gamma, family with sequence similarity 120B, FLJ55614, KIAA1838constitutive coactivator of peroxisome proliferator-activated receptor gamma, PGCC1dJ894D12.1, PPARG constitutive coactivator 1, PPARgamma constitutive coactivator 1, Protein FAM120B | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This antibody will not cross-react with FAM120A. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 0.5-1 μg/mL, ELISA 1:100-1:2000 | |
| Q96EK7 | |
| FAM120B | |
| Antibody was raised against an 18 amino acid synthetic peptide from near the carboxy terminus of human PGCC1. The immunogen is located within the last 50 amino acids of PGCC1. Amino Acid Squence: RDQGPGSRQYEHDQWRRY | |
| 0.025 mg | |
| Lipid and Metabolism | |
| 84498 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction