missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM120B Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76315MFV500
This item is not returnable.
View return policy
Description
FAM120B Polyclonal antibody specifically detects FAM120B in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
Specifications
| FAM120B | |
| Polyclonal | |
| Western Blot, ELISA | |
| CCPGKIAA0183, Constitutive coactivator of PPARG, Constitutive coactivator of PPAR-gamma, family with sequence similarity 120B, FLJ55614, KIAA1838constitutive coactivator of peroxisome proliferator-activated receptor gamma, PGCC1dJ894D12.1, PPARG constitutive coactivator 1, PPARgamma constitutive coactivator 1, Protein FAM120B | |
| Antibody was raised against an 18 amino acid synthetic peptide from near the carboxy terminus of human PGCC1. The immunogen is located within the last 50 amino acids of PGCC1. Amino Acid Squence: RDQGPGSRQYEHDQWRRY | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 84498 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction