missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM122C Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569857
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 83%; Dog: 92%; Guinea Pig: 83%; Horse: 83%; Human: 100%; Mouse: 83%; Pig: 85%; Rabbit: 83%; Rat: 83%.
FAM122C is a protein coding gene.
Specifications
| FAM122C | |
| Polyclonal | |
| Unconjugated | |
| FAM122C | |
| 4930432H15Rik; 4930527G05Rik; FAM122C; family with sequence similarity 122, member C; family with sequence similarity 122C; protein FAM122C; uncharacterized protein LOC685701 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 159091 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q6P4D5 | |
| FAM122C | |
| synthetic peptide directed towards the N-terminal region of human FAM122C. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction