missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM131C Monoclonal Antibody (OTI6G2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810783S
This item is not returnable.
View return policy
Description
FAM131C Monoclonal antibody specifically detects FAM131C in Human samples. It is validated for Western BlotSpecifications
| FAM131C | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9BUH8 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 57596 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI6G2 | |
| Unconjugated | |
| BEGAIN | |
| C1orf117; FAM131C; family with sequence similarity 131 member C; family with sequence similarity 131, member C; Gm693; hypothetical protein LOC619198; protein FAM131C; RP11-5P18.9; zgc:110264 | |
| Human recombinant protein fragment corresponding to amino acids 374-517 of human BEGAIN produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction