Learn More
FAM134A Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24650-1-AP-20UL
Description
The FAM134A antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human FAM134A. This antibody recognizes human, mouse, rat antigen. The FAM134A antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| FAM134A | |
| 550 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |
| Western Blot, Immunofluorescence, Immunohistochemistry, ELISA | |
| C2orf17, FAM134A, Protein FAM134A | |
| 20 μL | |
| 79137 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |