Learn More
FAM134A Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 24650-1-AP-20UL
 View more versions of this product
Product Code. 18080456
  • € 145.00 / 20µL
Description
The FAM134A antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human FAM134A. This antibody recognizes human, mouse, rat antigen. The FAM134A antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
FAM134A
550 μg/mL
Rabbit
Primary
Human, Mouse, Rat
150 kDa
IgG
Western Blot, Immunofluorescence, Immunohistochemistry, ELISA
C2orf17, FAM134A, Protein FAM134A
20 μL
79137
Store at -20°C. Stable for one year after shipment.