missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM14A Polyclonal Antibody

Beskrivelse
This target displays homology in the following species: Cow: 77%; Human: 100%.
IFI27L2 plays a role in the apoptotic process and has a pro-apoptotic activity.
Tekniske data
Tekniske data
| Antigen | FAM14A |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | IFI27L2 |
| Gene Accession No. | Q9H2X8 |
| Gene Alias | FAM14A; family with sequence similarity 14, member A; IFI27L2; interferon alpha inducible protein 27 like 2; interferon alpha-inducible protein 27-like protein 2; interferon, alpha-inducible protein 27-like 2; interferon-stimulated gene 12b protein; ISG12(b); ISG12(b) protein; ISG12B; pIFI27-like protein; Protein TLH29; TLH29; Unknown (protein for MGC:140204) |
| Gene Symbols | IFI27L2 |
| Vis mere |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?