missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FAM155A Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA558792

Product Code. 15776661

  • € 598.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: GNRGKDDRGK ALFLGNSAKP VWRLETCYPQ GASSGQCFTV ENADAVCARN WSRGAAGGDG QEVRSKHPTP L Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 28%.

FAM155A gene ontology annotations related to this gene include calcium ion import across plasma membrane.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FAM155A
Polyclonal
Unconjugated
FAM155A
6430500D16; AW121567; Fam155a; family with sequence similarity 155 member A; family with sequence similarity 155, member A; RGD1559498; Tmem28; transmembrane protein 28; transmembrane protein FAM155A
Rabbit
Antigen affinity chromatography
RUO
728215
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
B1AL88
FAM155A
Recombinant protein corresponding to Human FAM155A. Recombinant protein control fragment (Product #RP-98014).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.