missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM164A Monoclonal Antibody (OTI4C1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA505191S
This item is not returnable.
View return policy
Description
FAM164A Monoclonal antibody specifically detects FAM164A in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western BlotSpecifications
| FAM164A | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q96GY0 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 51101 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI4C1 | |
| Unconjugated | |
| ZC2HC1A | |
| 3110050N22Rik; AI790358; AU023959; C14H8ORF70; C8orf70; CGI-62; Fam164a; family with sequence similarity 164, member A; protein FAM164A; RGD1311970; UPF0418 protein FAM164A; Zc2hc1a; Zinc finger C2HC domain-containing protein 1A; zinc finger C2HC-type containing 1A; zinc finger, C2HC-type containing 1A | |
| Full length human recombinant protein of human FAM164A produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction