missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM170A Monoclonal Antibody (OTI6E4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809624S
This item is not returnable.
View return policy
Description
FAM170A Monoclonal antibody specifically detects FAM170A in Human samples. It is validated for Western BlotSpecifications
| FAM170A | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O00470 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 4211 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI6E4 | |
| Unconjugated | |
| MEIS1 | |
| FAM170A; family with sequence similarity 170 member A; family with sequence similarity 170, member A; protein FAM170A; Zinc finger domain-containing protein; zinc finger protein ZNFD; Znf domain containing protein; ZNFD | |
| Human recombinant protein fragment corresponding to amino acids 176-390 of human MEIS1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction