missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM173A Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06308APC
This item is not returnable.
View return policy
Description
FAM173A Polyclonal antibody specifically detects FAM173A in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| FAM173A | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| family with sequence similarity 173, member A | |
| Produced in rabbits immunized with E. coli-derived Human FAM173A fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| APC | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 65990 | |
| Store at 4C in the dark. | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido