missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM173A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561198
This item is not returnable.
View return policy
Description
Immunogen sequence: CAGSVCYRRK DLWKVSLRDC RNVSVFLAPS VLPLLEDKLR TELPAGARVV SGRFPLPTWQ PVTAVGEGLD RVWAYDVPEG GQAGEAASSR IPIQAA Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 77%.
Specifications
| FAM173A | |
| Polyclonal | |
| Unconjugated | |
| FAM173A | |
| Adenine nucleotide translocase lysine N-methyltransferase; ANTKMT; ANT-KMT; C16orf24; FAM173A; family with sequence similarity 173 member A; family with sequence similarity 173, member A; protein FAM173A; RJD7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 65990 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BQD7 | |
| FAM173A | |
| Recombinant protein corresponding to Human FAM173A. Recombinant protein control fragment (Product #RP-98370). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction