missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FAM175B Polyclonal Antibody
GREENER_CHOICE

Product Code. 15795911
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15795911 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15795911 Supplier Invitrogen™ Supplier No. PA558027

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: DVEKSERVVE SCQAEVNKLR RQITQRKNEK EQERRLQQAV LSRQMPSESL DPAFSPRMPS SGFAAEGRST LGDA Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 92%.

Component of the BRISC complex, a multiprotein complex that specifically cleaves 'Lys-63'-linked polyubiquitin, leaving the last ubiquitin chain attached to its substrates (PubMed:19214193, PubMed:20032457, PubMed:20656690, PubMed:24075985). May act as a central scaffold protein that assembles the various components of the BRISC complex and retains them in the cytoplasm (PubMed:20656690). Plays a role in regulating the onset of apoptosis via its role in modulating 'Lys-63'-linked ubiquitination of target proteins. Required for normal mitotic spindle assembly and microtubule attachment to kinetochores via its role in deubiquitinating NUMA1 (PubMed:26195665). Plays a role in interferon signaling via its role in the deubiquitination of the interferon receptor IFNAR1; deubiquitination increases IFNAR1 activities by enhancing its stability and cell surface expression (PubMed:24075985, PubMed:26344097). Down-regulates the response to bacterial lipopolysaccharide (LPS) via its role in IFNAR1 deubiquitination (PubMed:24075985). Required for normal induction of p53/TP53 in response to DNA damage (PubMed:25283148). Independent of the BRISC complex, promotes interaction between USP7 and p53/TP53, and thereby promotes deubiquitination of p53/TP53, preventing its degradation and resulting in increased p53/TP53-mediated transcription regulation and p53/TP53-dependent apoptosis in response to DNA damage (PubMed:25283148). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM175B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ABRAXAS2
Gene Accession No. Q15018
Gene Alias AA589499; Abraxas brother; abraxas brother 1; abraxas brother protein 1; Abraxas2; Abro1; AI853413; BRISC complex subunit Abraxas 2; BRISC complex subunit Abro1; C430003P19Rik; Em:AC068896.4; FAM175B; family with sequence similarity 175 member B; family with sequence similarity 175, member B; KIAA0157; Protein FAM175B; RGD1308918
Gene Symbols ABRAXAS2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM175B. Recombinant protein control fragment (Product #RP-102776).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23172
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.