missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM177A1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563129
This item is not returnable.
View return policy
Description
Immunogen sequence: ESAGQMSNER GFENVELGVI GKKKKVPRRV IHFVSGETME EYSTDEDEVD GVEKKDV Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 86%.
This gene encodes a member of a conserved protein family. Alternative splicing results in multiple transcript variants. This gene is thought to be associated with susceptibility to juvenile idiopathic arthritis.
Specifications
| FAM177A1 | |
| Polyclonal | |
| Unconjugated | |
| FAM177A1 | |
| C14orf24; FAM177A1; family with sequence similarity 177 member A1; family with sequence similarity 177, member A1; protein FAM177A1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 283635 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8N128 | |
| FAM177A1 | |
| Recombinant protein corresponding to Human FAM177A1. Recombinant protein control fragment (Product #RP-101371). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction