missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM198A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561576
This item is not returnable.
View return policy
Description
Immunogen sequence: TSGLTLWPHT AEGRDLLGAE NRALTGGQQA EDPTLASGAH QWPGSVEKLQ GSVWCDAETL LSSSRTGGQA PPWLTDHDVQ MLR Highest antigen sequence identity to the following orthologs: Mouse - 46%, Rat - 45%.
Specifications
| FAM198A | |
| Polyclonal | |
| Unconjugated | |
| FAM198A | |
| C3orf41; FAM198A; family with sequence similarity 198 member A; family with sequence similarity 198, member A; GASK1A; Golgi-associated kinase 1A; Protein FAM198A | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 729085 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UFP1 | |
| FAM198A | |
| Recombinant protein corresponding to Human FAM198A. Recombinant protein control fragment (Product #RP-100152). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction