missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM205A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 369.00
Specifications
| Antigen | FAM205A |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:500 - 1:1000 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
FAM205A Polyclonal specifically detects FAM205A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| FAM205A | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 259308 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:EDSMFSAAAKVAKTRKENVAKSLAPAKSPVGRSKTEKPTGCSKAQSRPAQKLVGPAFLDGPQSLDDKLRLHSRQPGSASALGYPRHCPRHCPREACANKPGHPT | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| C9orf144B, family with sequence similarity 205, member A, RP11-195F19.10 | |
| FAM205A | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title