missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM213A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-48517
This item is not returnable.
View return policy
Description
FAM213A Polyclonal antibody specifically detects FAM213A in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| FAM213A | |
| Polyclonal | |
| Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| C10orf58, Chromosome 10 Open Reading Frame 58, Family With Sequence Similarity 213, Member A, PAMM, Peroxiredoxin (PRX)-Like 2 Activated In M-CSF Stimulated Monocytes, Peroxiredoxin-Like 2 Activated In M-CSF Stimulated Monocytes, Protein PAMM, Redox-Regulatory Protein PAMM, UPF0765 Protein C10orf58 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: NTDVFLSKPQKAALEYLEDIDLKTLEKEPRTFKAKELWEKNGAVIMAVRRPGCFLCREEAADLSSLKSMLDQLGVPLYAVVKEHIRTEVKDFQPYFKGE | |
| 0.1 mL | |
| Primary | |
| Human, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 84293 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction