missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM213B Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98197AF405
This item is not returnable.
View return policy
Description
FAM213B Polyclonal antibody specifically detects FAM213B in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| FAM213B | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 127281 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Alexa Fluor 405 | |
| 2810405K02Rik, C1orf93, chromosome 1 open reading frame 93, DKFZp547M123, family with sequence similarity 213, member B, prostamide/PG F synthase | |
| Produced in rabbits immunized with E. coli-derived Human FAM213B fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction