missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM25A Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560835
This item is not returnable.
View return policy
Description
Immunogen sequence: EVVGHAKETG EKAIAEAIKK AQESGDKKMK EITETVTNTV TNAITHAAES L Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 71%.
FAM25A is a protein coding gene.
Specifications
| FAM25A | |
| Polyclonal | |
| Unconjugated | |
| FAM25A | |
| bA96C23.5; FAM25A; FAM25C; FAM25G; family with sequence similarity 25 member A; family with sequence similarity 25, member A; protein FAM25; protein FAM25A; Protein FAM25C; Protein FAM25G | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 643161 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| B3EWG3 | |
| FAM25A | |
| Recombinant protein corresponding to Human FAM25B. Recombinant protein control fragment (Product #RP-104373). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction