missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM3A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569619
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 85%.
A novel cytokine-like gene family using structure-based methods to search for novel four-helix-bundle cytokines in genomics databases. There are four genes in this family, FAM3A, FAM3B, FAM3C, and FAM3D, each encoding a protein (224-235 amino acids) with a hydrophobic leader sequence. Northern analysis indicates that FAM3B is highly expressed in pancreas, FAM3D in placenta, and FAM3A and FAM3C in almost all tissues. Immunohistochemistry showed that FAM3A is expressed prominently in the vascular endothelium, particularly capillaries. Fam3a staining in capillary endothelium and endocardium of heart, as well as vascular endothelial staining in many other tissues.
Specifications
| FAM3A | |
| Polyclonal | |
| Unconjugated | |
| FAM3A | |
| 1810037C20Rik; 2.19; 2-19; Cytokine-like protein 2-19; DLD; DXS560S; FAM3 metabolism regulating signaling molecule A; FAM3A; family 3, member A protein; family with sequence similarity 3 member A; family with sequence similarity 3, member A; hypothetical protein MGC73350; protein FAM3A; RGD1562076; XAP-7; zgc:73350 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 501664 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| 0 | |
| FAM3A | |
| synthetic peptide directed towards the C-terminal region of rat FAM3A. | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction