missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM3D Monoclonal Antibody (K29027_2D4), Invitrogen™
Mouse Monoclonal Antibody
€ 480.00 - € 846.00
Specifications
| Antigen | FAM3D |
|---|---|
| Clone | K29027_2D4 |
| Concentration | 0.98 mg/mL |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30204755
|
Invitrogen™
MA556186 |
200 μL |
€ 846.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30205435
|
Invitrogen™
MA555806 |
50 μL |
€ 480.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: YMSFSMKTIR LPRWLAASPT KEIQVKKYKC GLIKPCPANY FAFKICSGAA NVVGPTMCFE DRMIMSPVKN NVGRGLNIAL VNGTTGAVLG QKAFDMYSGD VMHLVKFLKE IPGGALVLVA SYDDPGTKMN DESRKLFSDL GSSYAKQLGF RDSWVFIGAK DLRGKSPFEQ FLKNSPDTNK YEGWPELLEM EGCMPPKPF
FAM3D Monoclonal antibody specifically detects FAM3D in Human samples. It is validated for Immunoprecipitation, Western BlotSpecifications
| FAM3D | |
| 0.98 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| 2310076N21Rik; AV067083; cytokine-like protein EF-7; EF7; EF-7; Fam3d; family with sequence similarity 3 member D; family with sequence similarity 3, member D; Oit1; oncoprotein induced transcript 1; oncoprotein induced transcript 1 homolog; oncoprotein-induced protein 1; Protein EF-7; protein FAM3D; UNQ567/PRO1130 | |
| Recombinant human FAM3D (immunogen range is 26-224). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Fam3d |
| K29027_2D4 | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q96BQ1 | |
| 131177 | |
| IgG1 | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title