missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM44A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA540107
This item is not returnable.
View return policy
Description
Recommended positive controls: SH-SY5Y nuclear extract. Predicted reactivity: Bovine (81%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The FAM44A gene, also known as Family With Sequence Similarity 44 Member A, is located on chromosome 10q11.22. This gene encodes a protein that is implicated in cellular processes but its precise biological function remains to be fully elucidated. FAM44A's expression is observed predominantly in the testes, and it is believed to potentially play a role in spermatogenesis. Structurally, it includes various exons and introns characteristic of protein-coding genes. FAM44A is considered part of poorly characterized genes, and ongoing research is aimed at understanding its functional dynamics, potential roles in disease conditions, and interactions with other proteins. Databases like STRING suggest protein-protein interactions involving FAM44A, indicating its participation in complex cellular networks.
Specifications
| FAM44A | |
| Polyclonal | |
| Unconjugated | |
| BOD1L1 | |
| biorientation of chromosomes in cell division 1 like 1; biorientation of chromosomes in cell division 1-like 1; biorientation of chromosomes in cell division protein 1-like 1; BOD1L; BOD1L1; FAM44A; family with sequence similarity 44, member A; KIAA1327 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 259282 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1.01 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% Thimerosal; pH 7 | |
| Q8NFC6 | |
| BOD1L1 | |
| Recombinant protein encompassing a sequence within the center region of human FAM44A. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction