missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM44B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550632
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total BOD1 protein.
FAM44B required for proper chromosome biorientation through the detection or correction of syntelic attachments in mitotic spindles.
Specifications
| FAM44B | |
| Polyclonal | |
| Unconjugated | |
| BOD1 | |
| 2310022M17Rik; biorientation defective 1; biorientation defective protein 1; biorientation of chromosomes in cell division 1; biorientation of chromosomes in cell division protein 1; BOD1; BOD2.1; bone length and organs 1; Fam44b; family with sequence similarity 44, member B; Protein FAM44B; RGD1305622 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 91272 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1.6 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q96IK1 | |
| BOD1 | |
| Synthetic peptide corresponding to residues near the C terminal of human biorientation of chromosomes in cell division 1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction