missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FAM45A Polyclonal Antibody
GREENER_CHOICE

Product Code. 18025791
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18025791 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18025791 Supplier Invitrogen™ Supplier No. PA5144897

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: SIKDIVSQFG METVILHTAL MLKKRIVVYH PKIEAVQEFT RTLPALVWHR QDWTILHSYV HLNADELEA Antigen sequence identity: Mouse 94%; Rat 91%.

FAM45A is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM45A
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM45A
Gene Accession No. Q8TCE6
Gene Alias DENN domain-containing protein 10; DENND10; FAM45A; family with sequence similarity 45 member A; family with sequence similarity 45, member A; protein FAM45A
Gene Symbols FAM45A
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 158-226. Recombinant protein control fragment (Product #RP-110007).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 404636
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.