missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM45A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA571395
This item is not returnable.
View return policy
Description
FAM45A Polyclonal Antibody for Western Blot
FAM45A is a protein coding gene.
Specifications
| FAM45A | |
| Polyclonal | |
| Unconjugated | |
| FAM45A | |
| DENN domain-containing protein 10; DENND10; FAM45A; family with sequence similarity 45 member A; family with sequence similarity 45, member A; protein FAM45A | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 404636 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8TCE6 | |
| FAM45A | |
| synthetic peptide directed towards the middle region of human FAM45A. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction