missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FAM47A Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA569965

Product Code. 15858373

  • € 530.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This target displays homology in the following species: Human: 100%.

FAM47A is a protein coding gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FAM47A
Polyclonal
Unconjugated
FAM47A
FAM47A; family with sequence similarity 47 member A; family with sequence similarity 47, member A; protein FAM47A
Rabbit
Affinity chromatography
RUO
158724
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q5JRC9
FAM47A
synthetic peptide directed towards the middle region of human FAM47A.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.