missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FAM47E Polyclonal Antibody
GREENER_CHOICE

Product Code. 15790562 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15790562 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15790562 Supplier Invitrogen™ Supplier No. PA562777

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: PHKMDLLHEN GPRPGLHENV CKAVSDFCKW VTTFGISDID EEFILKQFDI DYETKPSHDA LHTMKLNQVP LELKRSVGLS K Highest antigen sequence identity to the following orthologs: Mouse - 38%, Rat - 38%.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM47E
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM47E
Gene Accession No. Q6ZV65
Gene Alias C730043O17; FAM47E; family with sequence similarity 47 member E; family with sequence similarity 47, member E; Gm1381; Protein FAM47E
Gene Symbols FAM47E
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM47E. Recombinant protein control fragment (Product #RP-100286).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100129583
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.