missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM50B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578468
This item is not returnable.
View return policy
Description
Positive Control: 293T, MCF-7, MDA-MB-231 Predicted Reactivity: Mouse (89%), Rat (90%), Rhesus Monkey (98%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
FAM50B contains an intronless ORF that arose from ancestral retroposition. The encoded protein is related to a plant protein that plays a role in the circadian clock. FAM50B is adjacent to a differentially methylated region (DMR) and is imprinted and paternally expressed in many tissues.
Specifications
| FAM50B | |
| Polyclonal | |
| Unconjugated | |
| FAM50B | |
| D0H6S2654E; D6S2654E; FAM50B; family with sequence similarity 50 member B; family with sequence similarity 50, member B; protein FAM50B; Protein XAP-5-like; RGD1563458; X5l; XAP-5-like; XAP5-like protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 26240 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1.0 mg/mL | |
| PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q9Y247 | |
| FAM50B | |
| Recombinant protein encompassing a sequence within the center region of human FAM50B. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction