missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FAM53A Polyclonal Antibody

Product Code. 15761302 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15761302 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15761302 Supplier Invitrogen™ Supplier No. PA563529

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: PCDSRGLPGI TMPGCSQRGL RTSPVHPNLW ASRESVTSDG SRRSSGDPRD GDSVGEEGVF PRARWELDLE QIENN Highest antigen sequence identity to the following orthologs: Mouse - 55%, Rat - 56%.

Dorsal neural-tube nuclear protein is a 398 amino acid nuclear protein. DNTNP is thought to play an important role in neural development by specifying dorsal cell fates within the neural tube. Widely expressed in the dorsal neural tube, DNTNP is most highly expressed in the dorsal regions of the midbrain, the hindbrain, the diencephalon, and the spinal neural tube and is expressed at lower levels in the branchial arches, the telencephalon, the heart, and somites of developing embryos. DNTNP is encoded by a gene located on chromosome 4 which contains many genes including the Huntingtin gene, which is found to encode an expanded glutamine tract in cases of Huntington's disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM53A
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM53A
Gene Accession No. Q6NSI3
Gene Alias DNTNP; Dorsal neural-tube nuclear protein; FAM53A; family with sequence similarity 53 member A; family with sequence similarity 53, member A; protein FAM53A
Gene Symbols FAM53A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM53A. Recombinant protein control fragment (Product #RP-103747).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 152877
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.