missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM54A Monoclonal Antibody (OTI2G8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811420S
This item is not returnable.
View return policy
Description
FAM54A Monoclonal antibody specifically detects FAM54A in Human samples. It is validated for Western BlotSpecifications
| FAM54A | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q2M3W8 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 339318 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI2G8 | |
| Unconjugated | |
| ZNF181 | |
| DUF729 domain containing 1; DUF729 domain-containing protein 1; DUFD1; EGK_15321; FAM54A; family with sequence similarity 54, member A; LOC100911069; Mitochondrial fission regulator 2; Mtfr2; protein FAM54A; protein FAM54A-like | |
| Full length human recombinant protein of human ZNF181 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction