missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM54A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA543741
This item is not returnable.
View return policy
Description
Peptide sequence: NKTNYSHHSK SQRNKDIPNM LDVLKDMNKV KLRAIERSPG GRPIHKRKRQ Sequence homology: Cow: 85%; Human: 100%; Rabbit: 83%.
FAM54A belongs to the MTFR1/FAM54 family. The function of the FAM54A protein is not known.
Specifications
| FAM54A | |
| Polyclonal | |
| Unconjugated | |
| MTFR2 | |
| DUF729 domain containing 1; DUF729 domain-containing protein 1; DUFD1; EGK_15321; FAM54A; family with sequence similarity 54, member A; LOC100911069; Mitochondrial fission regulator 2; Mtfr2; protein FAM54A; protein FAM54A-like | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 113115 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q6P444 | |
| MTFR2 | |
| Synthetic peptide directed towards the middle region of human FAM54A. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction