missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM55B Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00120406-B01P-50ug
This item is not returnable.
View return policy
Description
FAM55B Polyclonal antibody specifically detects FAM55B in Human samples. It is validated for Western Blot
Specifications
| FAM55B | |
| Polyclonal | |
| PBS (pH 7.4) | |
| Family With Sequence Similarity 55, Member B, FLJ25224, Neurexophilin And PC-Esterase Domain Family, Member 2, NXPE Family Member 2, NXPE2, Protein FAM55B | |
| FAM55B (AAI56725.1, 1 a.a. - 365 a.a.) full-length human protein. MVEKILIHRILTLFPNAIARKLLLMLTFILIFWIIYLASKDHTKFSFNLENHIILNQGNIFKKYSHSETPLCPAVSPKETELRIKDIMEKLDQQIPPRPFTHVNTTTSATHSTATILNPQDTYCRGDQLDILLEVRDHLGHRKQYGGDFLRARMYSTALMAGASGKVTDFNNGTYLVSFTLFWEGQVSLSLLLIHPSEGVSALWRARNQGCDRIIFTGLFANRSSNVFTECGLTLNTNAELCQYMDDRDQEAFYCVRPQHMPCEALTHMTTRTRNISYLSKEEWRLFHRSNIGVEMMKNFTPIEVIPCPALFYFIFRDSLTLSPRLECSGMISAHCNLCLPGSSDSPDSASHVAGITSVQRHTWL | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| AAI56725.1 | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 120406 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction