missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM59B Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41118IR
This item is not returnable.
View return policy
Description
FAM59B Polyclonal antibody specifically detects FAM59B in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| FAM59B | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 755 | |
| FAM59B, Family With Sequence Similarity 59, Member B, GAREML, GRB2 Associated, Regulator Of MAPK1-Like, GRB2-Associated And Regulator Of MAPK Protein-Like, GRB2-Associated And Regulator Of MAPK1-Like, KIAA2038, Protein FAM59B | |
| Antibody was raised against a 19 amino acid synthetic peptide from near the carboxy terminus of human FAM59B. The immunogen is located within amino acids 600 - 650 of FAM59B. Amino Acid Squence: PPLFKPSHPQKRFAPFGA | |
| 0.1 mL | |
| Signal Transduction | |
| 150946 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction