Learn More
FAM64A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand:  Novus Biologicals NBP2-14002
 View more versions of this product
Product Code. 18700634
  • ONLINE EXCLUSIVE PRICE
    € 593.00
    € 560.70 / 0.10mL
    Save € 32.30
    5% Off
Description
FAM64A Polyclonal specifically detects FAM64A in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FAM64A