missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM65B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104330
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total FAM65B.
Required for hearing (PubMed:24958875). Involved in skeletal muscle development (PubMed:24687993).
Specifications
| FAM65B | |
| Polyclonal | |
| Unconjugated | |
| Ripor2 | |
| Ab2-162; C6orf32; DFNB104; DIFF40; DIFF48; FAM65B; family with sequence similarity 65 member B; family with sequence similarity 65, member B; Kiaa0386; myogenesis-related and NCAM-associated protein homolog; MYONAP; PL48; protein FAM65B; RGD1306939; Rho family-interacting cell polarization regulator 2; RIPOR2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 193385, 306934, 9750 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q7TP54, Q80U16, Q9Y4F9 | |
| Ripor2 | |
| A synthesized peptide derived from human RIPOR2(Accession Q9Y4F9), corresponding to amino acid residues E312-L362. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction